Skip to Content
Merck

BDPH9BCDBEA6

Bide Pharmatech

2-(3-Bromophenyl)acetamide

Synonym(s):

2-(3-bromophenyl)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H8O1N1Br1
CAS Number:
Molecular Weight:
214.06
MDL number:
Product BDPH9BCDBEA6 is not currently sold in your country.


Inchi Code

1S/C8H8BrNO/c9-7-3-1-2-6(4-7)5-8(10)11/h1-4H,5H2,(H2,10,11)

Inchi Key

JEYSSRJRKIAJIO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2-(3-Bromophenyl)acetamide


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library