Skip to Content
Merck

BDPH9BCDBF36

Bide Pharmatech

IV-361

Synonym(s):

N-(5-fluoro-2-methylphenyl)-6,6-dimethyl-3-(1-(trimethylsilyl)cyclobutane-1-carboxamido)-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C23H32O2N5F1Si1
CAS Number:
Molecular Weight:
457.62
MDL number:
Product BDPH9BCDBF36 is not currently sold in your country.


Inchi Code

1S/C23H32FN5O2Si/c1-14-8-9-15(24)12-17(14)25-21(31)29-13-16-18(22(29,2)3)27-28-19(16)26-20(30)23(10-7-11-23)32(4,5)6/h8-9,12H,7,10-11,13H2,1-6H3,(H,25,31)(H2,26,27,28,30)

Inchi Key

FYIUNJAFLWNBAI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

99%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under IV-361



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library