Skip to Content

BDPH9BCDC2E6

Bide Pharmatech

3,5-Dichloro-1H-1,2,4-triazole

Synonym(s):

3,5-dichloro-1H-1,2,4-triazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C2H1N3Cl2
CAS Number:
Molecular Weight:
137.96
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C2HCl2N3/c3-1-5-2(4)7-6-1/h(H,5,6,7)

Inchi Key

JNTFFNKMQMJJQZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 3,5-Dichloro-1H-1,2,4-triazole


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library