Skip to Content

BDPH9BCDD0FA

Bide Pharmatech

Methyl 2-[4-(trifluoromethyl)phenyl]acetate

Synonym(s):

methyl [4-(trifluoromethyl)phenyl]acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H9O2F3
CAS Number:
Molecular Weight:
218.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H9F3O2/c1-15-9(14)6-7-2-4-8(5-3-7)10(11,12)13/h2-5H,6H2,1H3

Inchi Key

ONNMKZXKTBYWFA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid or Semi-solid or liquid or lump



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library