Skip to Content
Merck

BDPH9BCDD87A

Bide Pharmatech

8-Methyl-1-naphthoic acid

Synonym(s):

8-methyl-1-naphthoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H10O2
CAS Number:
Molecular Weight:
186.21
MDL number:
Product BDPH9BCDD87A is not currently sold in your country.


Inchi Code

1S/C12H10O2/c1-8-4-2-5-9-6-3-7-10(11(8)9)12(13)14/h2-7H,1H3,(H,13,14)

Inchi Key

FLBHUCKQJRXXQB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 8-Methyl-1-naphthoic acid


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library