Skip to Content
Merck

BDPH9BCDDBC8

Bide Pharmatech

Methyl 3-bromo-1-oxo-1H-indene-5-carboxylate

Synonym(s):

methyl 3-bromo-1-oxo-1H-indene-5-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H7O3Br1
CAS Number:
Molecular Weight:
267.08
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H7BrO3/c1-15-11(14)6-2-3-7-8(4-6)9(12)5-10(7)13/h2-5H,1H3

Inchi Key

WPVUELSDBGETTO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library