Skip to Content
Merck

BDPH9BCDDDF9

Bide Pharmatech

2-Fluoro-3-isobutoxy-4-methoxyphenylboronic acid

Synonym(s):

2-fluoro-3-isobutoxy-4-methoxyphenylboronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H16O4F1B1
CAS Number:
Molecular Weight:
242.06
MDL number:
Product BDPH9BCDDDF9 is not currently sold in your country.


Inchi Code

1S/C11H16BFO4/c1-7(2)6-17-11-9(16-3)5-4-8(10(11)13)12(14)15/h4-5,7,14-15H,6H2,1-3H3

Inchi Key

CVTXHALYOKFYGH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library