Skip to Content

BDPH9BCDE135

Bide Pharmatech

4-Cyclopropyl-1H-1,2,3-triazole-5-carbaldehyde

Synonym(s):

4-cyclopropyl-1H-1,2,3-triazole-5-carbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H7N3O/c10-3-5-6(4-1-2-4)8-9-7-5/h3-4H,1-2H2,(H,7,8,9)

Inchi Key

ZVGKPWQHQQAZGT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library