Skip to Content
Merck

BDPH9BCDE24D

Bide Pharmatech

5-Fluoro-2-methyl-1H-pyrrolo[2,3-b]pyridine

Synonym(s):

5-fluoro-2-methyl-1H-pyrrolo[2,3-b]pyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H7N2F1
CAS Number:
Molecular Weight:
150.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H7FN2/c1-5-2-6-3-7(9)4-10-8(6)11-5/h2-4H,1H3,(H,10,11)

Inchi Key

RSYDPOVVRMCJHZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library