Skip to Content
Merck

BDPH9BCDED7B

Bide Pharmatech

2-Chloro-N-methyl-N-(1,2,3,4-tetrahydronaphthalen-1-yl)acetamide

Synonym(s):

2-chloro-N-methyl-N-(1,2,3,4-tetrahydro-1-naphthalenyl)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H16O1N1Cl1
CAS Number:
Molecular Weight:
237.73
MDL number:
Product BDPH9BCDED7B is not currently sold in your country.


Inchi Code

1S/C13H16ClNO/c1-15(13(16)9-14)12-8-4-6-10-5-2-3-7-11(10)12/h2-3,5,7,12H,4,6,8-9H2,1H3

Inchi Key

FLTYMFJRZLOGIY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library