Skip to Content
Merck

BDPH9BCDEFE8

Bide Pharmatech

2-Chloro-5-(prop-1-en-2-yl)thiophene

Synonym(s):

2-chloro-5-isopropenylthiophene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H7Cl1S1
CAS Number:
Molecular Weight:
158.65
MDL number:
Product BDPH9BCDEFE8 is not currently sold in your country.


Inchi Code

1S/C7H7ClS/c1-5(2)6-3-4-7(8)9-6/h3-4H,1H2,2H3

Inchi Key

IUFGDQVHPWPKKZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library