Skip to Content
Merck

BDPH9BCDF672

Bide Pharmatech

DBCO-PEG4-triethoxysilane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C39H57O10N3Si1
CAS Number:
Molecular Weight:
755.98
MDL number:
Product BDPH9BCDF672 is not currently sold in your country.


Inchi Code

1S/C39H57N3O10Si/c1-4-50-53(51-5-2,52-6-3)31-11-21-40-38(44)20-23-46-25-27-48-29-30-49-28-26-47-24-22-41-37(43)18-19-39(45)42-32-35-14-8-7-12-33(35)16-17-34-13-9-10-15-36(34)42/h7-10,12-15H,4-6,11,18-32H2,1-3H3,(H,40,44)(H,41,43)

Inchi Key

BVPATRYMIHMPFW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?

Explore all of our products under DBCO-PEG4-triethoxysilane



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library