Skip to Content
Merck

BDPH9BCDF7C3

Bide Pharmatech

5-Bromopyridine-3-thiol hydrochloride

Synonym(s):

5-bromopyridine-3-thiol hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H5N1Cl1Br1S1
CAS Number:
Molecular Weight:
226.52
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H4BrNS.ClH/c6-4-1-5(8)3-7-2-4;/h1-3,8H;1H

Inchi Key

JIYFPWOJOWONCN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library