Skip to Content
Merck

BDPH9BCDFBB5

Bide Pharmatech

2-(Aminomethyl)butan-1-ol

Synonym(s):

2-(aminomethyl)-1-butanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H13O1N1
CAS Number:
Molecular Weight:
103.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H13NO/c1-2-5(3-6)4-7/h5,7H,2-4,6H2,1H3

Inchi Key

DHPOUEPMENIWGX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 2-(Aminomethyl)butan-1-ol



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library