Skip to Content
Merck

BDPH9BCDFBF7

Bide Pharmatech

5-Methoxymethyl-thiophene

Synonym(s):

2-(methoxymethyl)thiophene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H8O1S1
CAS Number:
Molecular Weight:
128.19
MDL number:
Product BDPH9BCDFBF7 is not currently sold in your country.


Inchi Code

1S/C6H8OS/c1-7-5-6-3-2-4-8-6/h2-4H,5H2,1H3

Inchi Key

BMZQEYGEQALEIO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 5-Methoxymethyl-thiophene




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library