Skip to Content
Merck

BDPH9BCDFD34

Bide Pharmatech

6-Bromo-4-methylquinoline

Synonym(s):

6-bromo-4-methylquinoline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H8N1Br1
CAS Number:
Molecular Weight:
222.08
MDL number:
Product BDPH9BCDFD34 is not currently sold in your country.


Inchi Code

1S/C10H8BrN/c1-7-4-5-12-10-3-2-8(11)6-9(7)10/h2-6H,1H3

Inchi Key

XBZQSJLYRMZRSZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 6-Bromo-4-methylquinoline


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library