Skip to Content
Merck

BDPH9BCDFDE0

Bide Pharmatech

2-Bromo-4-(bromomethyl)thiazole

Synonym(s):

2-bromo-4-(bromomethyl)-1,3-thiazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H3N1Br2S1
CAS Number:
Molecular Weight:
256.95
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H3Br2NS/c5-1-3-2-8-4(6)7-3/h2H,1H2

Inchi Key

PUUHOIJJZAGFNK-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2-Bromo-4-(bromomethyl)thiazole



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library