Skip to Content
Merck

BDPH9BCDFF42

Bide Pharmatech

Tetrahydropyran-4-carbaldehyde

Synonym(s):

tetrahydro-2H-pyran-4-carbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H10O2
CAS Number:
Molecular Weight:
114.14
MDL number:
Product BDPH9BCDFF42 is not currently sold in your country.


Inchi Code

1S/C6H10O2/c7-5-6-1-3-8-4-2-6/h5-6H,1-4H2

Inchi Key

CXLGNJCMPWUZKM-UHFFFAOYSA-N

Shipping Temperature

Shipped in cold pack

Purity

95%

Country of Origin

CN

Physical Form

Liquid or semi-solid or solid

Storage temp.

Inert atmosphere, store in freezer, under -20°C



Still not finding the right product?

Explore all of our products under Tetrahydropyran-4-carbaldehyde




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library