Skip to Content
Merck

Skip To

BDPH9BCDFFA7

Bide Pharmatech

6-Chloro-5-iodonicotinic acid

Synonym(s):

6-chloro-5-iodonicotinic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C6H3O2N1Cl1I1
CAS Number:
Molecular Weight:
283.45
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H3ClINO2/c7-5-4(8)1-3(2-9-5)6(10)11/h1-2H,(H,10,11)

Inchi Key

PGYDTSFRXGYMNF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

White to yellow powder or crystals



Still not finding the right product?

Explore all of our products under 6-Chloro-5-iodonicotinic acid


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library