Skip to Content
Merck

BDPH9BCE062D

Bide Pharmatech

4-(4,6-Diphenyl-1,3,5-triazin-2-yl)aniline

Synonym(s):

4-(4,6-diphenyl-1,3,5-triazin-2-yl)aniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Product BDPH9BCE062D is not currently sold in your country.


Inchi Code

1S/C21H16N4/c22-18-13-11-17(12-14-18)21-24-19(15-7-3-1-4-8-15)23-20(25-21)16-9-5-2-6-10-16/h1-14H,22H2

Inchi Key

OFRZSDOXKFCVHD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library