Skip to Content
Merck

BDPH9BCE0DFE

Bide Pharmatech

Ethyl 5-bromo-6-oxo-1,6-dihydropyridazine-3-carboxylate

Synonym(s):

ethyl 5-bromo-6-hydroxypyridazine-3-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H7O3N2Br1
CAS Number:
Molecular Weight:
247.05
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H7BrN2O3/c1-2-13-7(12)5-3-4(8)6(11)10-9-5/h3H,2H2,1H3,(H,10,11)

Inchi Key

LRNXRVAADIJKCJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library