Skip to Content

BDPH9BCE0EE1

Bide Pharmatech

(3-(Cyclopropanecarbonyl)phenyl)boronic acid

Synonym(s):

3-(cyclopropylcarbonyl)phenylboronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H11O3B1
CAS Number:
Molecular Weight:
190.01
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H11BO3/c12-10(7-4-5-7)8-2-1-3-9(6-8)11(13)14/h1-3,6-7,13-14H,4-5H2

Inchi Key

WFAARFODOMXRPX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library