Skip to Content
Merck

BDPH9BCE1C5B

Bide Pharmatech

5-Amino-2-chloro-4-fluorobenzoic acid

Synonym(s):

5-amino-2-chloro-4-fluorobenzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H5O2N1Cl1F1
CAS Number:
Molecular Weight:
189.57
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H5ClFNO2/c8-4-2-5(9)6(10)1-3(4)7(11)12/h1-2H,10H2,(H,11,12)

Inchi Key

GANBUJGJERTPPV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library