Skip to Content
Merck

BDPH9C01ECBF

Bide Pharmatech

4-Amino-1-methyl-3,4-dihydroquinolin-2(1H)-one

Synonym(s):

4-amino-1-methyl-3,4-dihydro-2(1H)-quinolinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H12O1N2
CAS Number:
Molecular Weight:
176.22
MDL number:
Product BDPH9C01ECBF is not currently sold in your country.


Inchi Code

1S/C10H12N2O/c1-12-9-5-3-2-4-7(9)8(11)6-10(12)13/h2-5,8H,6,11H2,1H3

Inchi Key

FLIBAMYYJXENAJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library