Skip to Content
Merck

BDPH9C01EECF

Bide Pharmatech

2-Benzyl 1-(tert-butyl) (2S,4R)-4-methyl-5-oxopyrrolidine-1,2-dicarboxylate

Synonym(s):

2-benzyl 1-(tert-butyl) (2S,4R)-4-methyl-5-oxopyrrolidine-1,2-dicarboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Product BDPH9C01EECF is not currently sold in your country.


Inchi Code

1S/C18H23NO5/c1-12-10-14(16(21)23-11-13-8-6-5-7-9-13)19(15(12)20)17(22)24-18(2,3)4/h5-9,12,14H,10-11H2,1-4H3/t12-,14+/m1/s1

Inchi Key

SINJUUNBWYWVCW-OCCSQVGLSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library