Skip to Content
Merck

Skip To

BDPH9C01F1DD

Bide Pharmatech

(S)-3-Amino-4-methylpentan-1-ol hydrochloride

Synonym(s):

(S)-3-amino-4-methylpentan-1-ol hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C6H16O1N1Cl1
CAS Number:
Molecular Weight:
153.65
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H15NO.ClH/c1-5(2)6(7)3-4-8;/h5-6,8H,3-4,7H2,1-2H3;1H/t6-;/m0./s1

Inchi Key

SCSHCPKBDXKMIX-RGMNGODLSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library