Skip to Content

BDPH9C01F641

Bide Pharmatech

3-Bromo-1H-pyrrolo[3,2-c]pyridin-4-ol

Synonym(s):

3-bromo-1H-pyrrolo[3,2-c]pyridin-4-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H5O1N2Br1
CAS Number:
Molecular Weight:
213.03
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H5BrN2O/c8-4-3-10-5-1-2-9-7(11)6(4)5/h1-3,10H,(H,9,11)

Inchi Key

YJVORRLOJMKVSB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library