Skip to Content
Merck

BDPH9C0213AC

Bide Pharmatech

2,3-Dimethyl-1,5,6,7-tetrahydro-4H-indol-4-one

Synonym(s):

2,3-dimethyl-1,5,6,7-tetrahydro-4H-indol-4-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H13O1N1
CAS Number:
Molecular Weight:
163.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H13NO/c1-6-7(2)11-8-4-3-5-9(12)10(6)8/h11H,3-5H2,1-2H3

Inchi Key

UORPHFZBCZEBJV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library