Skip to Content
Merck

BDPH9C02370F

Bide Pharmatech

N-(3-Amino-3-oxoprop-1-en-2-yl)-2-((11S,14Z,21S,23S,29S)-14-ethylidene-3,23-dihydroxy-11-((1R)-(1-hydroxyethyl))-31-methyl-9,12,19,24,33,43-hexaoxo-9,10,11,12,13,14,19,20,21,22,23,24,26,33,35,36-hexadecahydro-21,36-([2,4]-endo-thiazolomethanimino)-5,8:15,18:37,40-tri(azeno)-30,32-epiminobenzo[h1]pyrido[3,2-v][1]oxa[10,20,26,30]tetrathia[6,13,16]triazacyclohexatriacontin-2-yl)thiazole-4-carboxamide

Synonym(s):

N-(3-amino-3-oxoprop-1-en-2-yl)-2-((5Z,14Z,15Z,31Z,37Z,45Z)-14-ethylidene-3,23-dihydroxy-11-(1-hydroxyethyl)-31-methyl-9,12,19,24,33,43-hexaoxo-9,10,11,12,13,14,19,20,21,22,23,24,26,33,35,36-hexadecahydro-21,36-([2,4]epithiazolomethanoimino)-5,8:15,18:37,40-tri(azeno)-30,32-epiminobenzo[h1]pyrido[3,2-v][1]oxa[10,20,26,30]tetrathia[6,13,16]triazacyclohexatriacontin-2-yl)thiazole-4-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C51H43O12N13S6
CAS Number:
Molecular Weight:
1222.38
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Storage temp.

Sealed in dry, store in freezer, under -20°C

Inchi Code

1S/C51H43N13O12S6/c1-5-23-46-60-28(14-79-46)41(70)56-25-10-33(67)50(74)76-11-21-7-6-8-24-34(21)18(2)35(54-24)51(75)82-17-31(57-42(71)29-15-80-47(25)61-29)48-58-26(12-78-48)37-22(45-59-30(13-77-45)43(72)64-36(20(4)65)44(73)55-23)9-32(66)38(63-37)49-62-27(16-81-49)40(69)53-19(3)39(52)68/h5-9,12-16,20,25,31,33,36,54,65-67H,3,10-11,17H2,1-2,4H3,(H2,52,68)(H,53,69)(H,55,73)(H,56,70)(H,57,71)(H,64,72)/b23-5-

Inchi Key

OQAOHXRUMXWDLQ-FPQMYIDKSA-N

Shipping Temperature

Normal

Purity

90%

Country of Origin

CN

Physical Form

Powder or crystals




hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library