Skip to Content
Merck

BDPH9C0238CC

Bide Pharmatech

Boc-Oxyma

Synonym(s):

ethyl (E)-2-(((tert-butoxycarbonyl)oxy)imino)-2-cyanoacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H14O5N2
CAS Number:
Molecular Weight:
242.23
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H14N2O5/c1-5-15-8(13)7(6-11)12-17-9(14)16-10(2,3)4/h5H2,1-4H3/b12-7+

Inchi Key

GTIPXWMOYWXFBG-KPKJPENVSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

White to pale-yellow solid



Still not finding the right product?

Explore all of our products under Boc-Oxyma


hstatements

Precautionary Statements

Signal Word

Danger

Pictograms

Flame


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library