Skip to Content
Merck

BDPH9C0244A8

Bide Pharmatech

4-Ethynyltetrahydro-2H-pyran-4-ol

Synonym(s):

4-ethynyltetrahydro-2H-pyran-4-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H10O2
CAS Number:
Molecular Weight:
126.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H10O2/c1-2-7(8)3-5-9-6-4-7/h1,8H,3-6H2

Inchi Key

SWJWVPMUAUALKX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

White to yellow solid



Still not finding the right product?

Explore all of our products under 4-Ethynyltetrahydro-2H-pyran-4-ol




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library