Skip to Content
Merck

BDPH9C02480E

Bide Pharmatech

Ethyl 4-(7-hydroxy-2,2,4-trimethylquinolin-1(2H)-yl)butanoate

Synonym(s):

ethyl 4-(7-hydroxy-2,2,4-trimethylquinolin-1(2H)-yl)butanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Product BDPH9C02480E is not currently sold in your country.


Inchi Code

1S/C18H25NO3/c1-5-22-17(21)7-6-10-19-16-11-14(20)8-9-15(16)13(2)12-18(19,3)4/h8-9,11-12,20H,5-7,10H2,1-4H3

Inchi Key

GPDUOVXIVXDETH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library