Skip to Content
Merck

Skip To

BL3H160B9783

BLD Pharmatech

Phthalic acid

Synonym(s):

phthalic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Ships in 5 days - from Aldrich Partner.
1 200,00 CZK
100 g
Ships in 5 days - from Aldrich Partner.
1 300,00 CZK
500 g
Ships in 5 days - from Aldrich Partner.
1 425,00 CZK
1 kg
Ships in 5 days - from Aldrich Partner.
2 200,00 CZK

About This Item

Linear Formula:
C8H6O4
CAS Number:
Molecular Weight:
166.13
MDL number:

Vendor SKU: BD147920-25G

1 200,00 CZK


Ships in 5 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H6O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4H,(H,9,10)(H,11,12)

Inchi Key

XNGIFLGASWRNHJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder Colour: White

Melting Point

191C



Still not finding the right product?

Explore all of our products under Phthalic acid


hstatements

Precautionary Statements

Signal Word

Danger

Pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library