Skip to Content
Merck

Skip To

BL3H160BB2E3

BLD Pharmatech

2-Bromo-5-nitrofuran

Synonym(s):

2-bromo-5-nitrofuran

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Ships in 10 days - from Aldrich Partner.
1 350,00 CZK
250 mg
Ships in 10 days - from Aldrich Partner.
2 100,00 CZK
1 g
Ships in 10 days - from Aldrich Partner.
4 200,00 CZK
5 g
Ships in 10 days - from Aldrich Partner.
11 525,00 CZK

About This Item

Linear Formula:
C4H2O3N1Br1
CAS Number:
Molecular Weight:
191.97
MDL number:

Vendor SKU: BD2183-100MG

1 350,00 CZK


Ships in 10 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H2BrNO3/c5-3-1-2-4(9-3)6(7)8/h1-2H

Inchi Key

QGTDMAKRJHGXOF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?

Explore all of our products under 2-Bromo-5-nitrofuran


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library