Skip to Content
Merck

BL3H160BCD2C

BLD Pharmatech

O-Acetyl-N-(tert-butyl)hydroxylamine hydrochloride

Synonym(s):

2-[(acetyloxy)amino]-2-methylpropane hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H14O2N1Cl1
CAS Number:
Molecular Weight:
167.64
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H13NO2.ClH/c1-5(8)9-7-6(2,3)4;/h7H,1-4H3;1H

Inchi Key

CWIOQGVXNNMFJC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Country of Origin

CN

Physical Form

White to Almost white powder to crystal

Storage temp.

Inert atmosphere, 2-8C



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library