Skip to Content
Merck

Skip To

BL3H160BE07A

BLD Pharmatech

4-Ethoxy-2-methylaniline

Synonym(s):

4-ethoxy-2-methylphenylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Ships in 10 days - from Aldrich Partner.
1 775,00 CZK
250 mg
Ships in 10 days - from Aldrich Partner.
2 400,00 CZK
1 g
Ships in 10 days - from Aldrich Partner.
5 750,00 CZK

About This Item

Linear Formula:
C9H13O1N1
CAS Number:
Molecular Weight:
151.21
MDL number:

Vendor SKU: BD283511-100MG

1 775,00 CZK


Ships in 10 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H13NO/c1-3-11-8-4-5-9(10)7(2)6-8/h4-6H,3,10H2,1-2H3

Inchi Key

GQMOFUZZCSQSNP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Liquid

Storage temp.

Keep in dark place, inert atmosphere, 2-8C



Still not finding the right product?

Explore all of our products under 4-Ethoxy-2-methylaniline


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library