Skip to Content
Merck

BL3H1F1CCBCC

BLD Pharmatech

5-Chlorothiophene-2-sulfonic acid tert-butylamide

Synonym(s):

N-(tert-butyl)-5-chloro-2-thiophenesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H12O2N1Cl1S2
CAS Number:
Molecular Weight:
253.77
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H12ClNO2S2/c1-8(2,3)10-14(11,12)7-5-4-6(9)13-7/h4-5,10H,1-3H3

Inchi Key

VPZSRWXWBIVADA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Off-white to orange powder or crystals



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library