Skip to Content
Merck

BL3H97A46D69

BLD Pharmatech

Bis(2-methylallyl)amine

Synonym(s):

2-methyl-N-(2-methyl-2-propenyl)-2-propen-1-amine

Select a Size

Change View

About This Item

Linear Formula:
C8H15N1
CAS Number:
Molecular Weight:
125.21
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H15N/c1-7(2)5-9-6-8(3)4/h9H,1,3,5-6H2,2,4H3

Inchi Key

LDCWGVLBCJEQMT-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Country of Origin

CN

Physical Form

Liquid

Storage temp.

Keep in dark place, sealed in dry, 2-8C



Still not finding the right product?

Explore all of our products under Bis(2-methylallyl)amine


hstatements

Precautionary Statements

Signal Word

Danger

Pictograms

FlameCorrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library