Skip to Content
Merck

BL3H97A46DF6

BLD Pharmatech

Ethyl 3-(2,6-dichlorophenyl)-3-oxopropanoate

Synonym(s):

ethyl 3-(2,6-dichlorophenyl)-3-oxopropanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H10O3Cl2
CAS Number:
Molecular Weight:
261.1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H10Cl2O3/c1-2-16-10(15)6-9(14)11-7(12)4-3-5-8(11)13/h3-5H,2,6H2,1H3

Inchi Key

GJUHMKHPXHMWQI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Solid or semi-solid or lump or liquid

Storage temp.

Sealed in dry, 2-8C



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library