Skip to Content
Merck

BL3H97A4969D

BLD Pharmatech

(3R,4R,4aR,6aR,6bS,8aR,11R,12S,12aR,14aR,14bS)-3-Acetoxy-4,6a,6b,8a,11,12,14b-heptamethyl-14-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,14,14a,14b-icosahydropicene-4-carboxylic acid

Synonym(s):

(3alpha,4alpha,9alpha)-3-(acetyloxy)-11-oxours-12-en-23-oic acid

Select a Size

Change View

About This Item

Linear Formula:
C32H48O5
CAS Number:
Molecular Weight:
512.73
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C32H48O5/c1-18-9-12-28(4)15-16-30(6)21(25(28)19(18)2)17-22(34)26-29(5)13-11-24(37-20(3)33)32(8,27(35)36)23(29)10-14-31(26,30)7/h17-19,23-26H,9-16H2,1-8H3,(H,35,36)/t18-,19+,23-,24-,25+,26-,28-,29+,30-,31-,32-/m1/s1

Inchi Key

HMMGKOVEOFBCAU-BCDBGHSCSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, store in freezer, under -20C

Country of Origin

CN

Physical Form

White to off-white solid




Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library