Skip to Content
Merck

BL3H97B9EC2D

BLD Pharmatech

Pyrimidine-2-carbothioamide

Synonym(s):

2-pyrimidinecarbothioamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H5N3S1
CAS Number:
Molecular Weight:
139.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H5N3S/c6-4(9)5-7-2-1-3-8-5/h1-3H,(H2,6,9)

Inchi Key

QAUHVPUYFSGVME-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to Yellow Powder or Crystals



hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library