Skip to Content
Merck

Skip To

BL3H97B9ECBB

BLD Pharmatech

4-(Methanesulfonyl)benzoyl chloride

Synonym(s):

4-(methylsulfonyl)benzoyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Ships in 5 days - from Aldrich Partner.
1 650,00 CZK
250 mg
Ships in 5 days - from Aldrich Partner.
2 150,00 CZK
1 g
Ships in 5 days - from Aldrich Partner.
4 375,00 CZK
5 g
Ships in 10 days - from Aldrich Partner.
13 775,00 CZK

About This Item

Linear Formula:
C8H7O3Cl1S1
CAS Number:
Molecular Weight:
218.66
MDL number:

Vendor SKU: BD308097-100MG

1 650,00 CZK


Ships in 5 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H7ClO3S/c1-13(11,12)7-4-2-6(3-5-7)8(9)10/h2-5H,1H3

Inchi Key

IPEIDGXNXFPGBG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library