Skip to Content
Merck

Skip To

BL3H97F030AA

BLD Pharmatech

2-(2-Methoxyethoxy)acetyl chloride

Synonym(s):

(2-methoxyethoxy)acetyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 mg
Ships in 5 days - from Aldrich Partner.
1 250,00 CZK
1 g
Ships in 10 days - from Aldrich Partner.
2 000,00 CZK
5 g
Ships in 10 days - from Aldrich Partner.
5 350,00 CZK
25 g
Ships in 10 days - from Aldrich Partner.
17 150,00 CZK

About This Item

Linear Formula:
C5H9O3Cl1
CAS Number:
Molecular Weight:
152.58
MDL number:

Vendor SKU: BD01155596-250MG

1 250,00 CZK


Ships in 5 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H9ClO3/c1-8-2-3-9-4-5(6)7/h2-4H2,1H3

Inchi Key

OZJZCCMIOZPPIT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Liquid

Storage temp.

Inert atmosphere, 2-8C



Still not finding the right product?

Explore all of our products under 2-(2-Methoxyethoxy)acetyl chloride



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library