Skip to Content
Merck

Skip To

BL3H98EDBD8A

BLD Pharmatech

(2S,3R)-3-Hydroxypyrrolidine-2-carboxylic acid hydrochloride

Synonym(s):

(2S,3R)-3-hydroxypyrrolidine-2-carboxylic acid hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mg
Ships in 10 days - from Aldrich Partner.
2 000,00 CZK
100 mg
Ships in 10 days - from Aldrich Partner.
2 300,00 CZK
250 mg
Ships in 10 days - from Aldrich Partner.
3 625,00 CZK
1 g
Ships in 10 days - from Aldrich Partner.
6 700,00 CZK
5 g
Ships in 10 days - from Aldrich Partner.
30 925,00 CZK
10 g
Ships in 10 days - from Aldrich Partner.
49 125,00 CZK

About This Item

Linear Formula:
C5H10O3N1Cl1
CAS Number:
Molecular Weight:
167.59
MDL number:

Vendor SKU: BD01313504-50MG

2 000,00 CZK


Ships in 10 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H9NO3.ClH/c7-3-1-2-6-4(3)5(8)9;/h3-4,6-7H,1-2H2,(H,8,9);1H/t3-,4+;/m1./s1

Inchi Key

HHFGQFQJUWOFCN-HJXLNUONSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Inert atmosphere, 2-8C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library