Skip to Content
Merck

BL3H98EDBF45

BLD Pharmatech

(1-Ethylpiperidin-4-yl)methanamine

Synonym(s):

(1-ethyl-4-piperidinyl)methanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H18N2
CAS Number:
Molecular Weight:
142.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Storage temp.

Keep in dark place, sealed in dry, 2-8C

Inchi Code

1S/C8H18N2/c1-2-10-5-3-8(7-9)4-6-10/h8H,2-7,9H2,1H3

Inchi Key

ZLODGCYXZYPQKQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under (1-Ethylpiperidin-4-yl)methanamine


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library