Skip to Content
Merck

BL3H9998EB01

BLD Pharmatech

2-(6-Bromo-1H-indol-3-yl)acetonitrile

Synonym(s):

(6-bromo-1H-indol-3-yl)acetonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H7N2Br1
CAS Number:
Molecular Weight:
235.08
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H7BrN2/c11-8-1-2-9-7(3-4-12)6-13-10(9)5-8/h1-2,5-6,13H,3H2

Inchi Key

MSIDSSLYDVKLRA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Inert atmosphere, 2-8C



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library