Storage temp.
Sealed in dry, 2-8C
Inchi Code
1S/C6H7F3O3/c7-6(8,9)12-4-1-3(2-4)5(10)11/h3-4H,1-2H2,(H,10,11)/t3-,4+
Inchi Key
MBYACEIXYASETJ-ZXZARUISSA-N
Shipping Temperature
Normal
Purity
95%
Country of Origin
CN
Physical Form
Solid or semi-solid or liquid
Still not finding the right product?
Explore all of our products under cis-3-(Trifluoromethoxy)cyclobutanecarboxylic acid
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
