Skip to Content
Merck

BL3H9BB32C92

BLD Pharmatech

4-Bromo-2-isopropyl-1H-imidazole

Synonym(s):

4-bromo-2-isopropyl-1H-imidazole

Select a Size

Change View

About This Item

Linear Formula:
C6H9N2Br1
CAS Number:
Molecular Weight:
189.05
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H9BrN2/c1-4(2)6-8-3-5(7)9-6/h3-4H,1-2H3,(H,8,9)

Inchi Key

QAXBJSJPKAAQRJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Powder or crystals

Storage temp.

Keep in dark place, inert atmosphere, 2-8C



Still not finding the right product?

Explore all of our products under 4-Bromo-2-isopropyl-1H-imidazole



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library