Skip to Content
Merck

BL3H9BB32D98

BLD Pharmatech

4-Bromo-3-fluoro-2-nitrophenol

Synonym(s):

4-bromo-3-fluoro-2-nitrophenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H3O3N1Br1F1
CAS Number:
Molecular Weight:
236
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H3BrFNO3/c7-3-1-2-4(10)6(5(3)8)9(11)12/h1-2,10H

Inchi Key

VQSACDGRYKNYRN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 4-Bromo-3-fluoro-2-nitrophenol



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library