Skip to Content
Merck

Skip To

D-205

Despropionyl meta-Fluorofentanyl solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

reference material

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H23FN2
CAS Number:
Molecular Weight:
298.40
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

application(s)

forensics and toxicology

format

single component solution

storage temp.

-10 to -25°C

General description

Despropionyl meta-Fluorofentany is a synthetic fentanyl analog which has potent analgesic properties. All three isomers, ortho-, meta-, and para- are emerging illicit drugs.

Application

Forensics and toxicology, clinical testing, cannabis, pharma, food and beverage, environmental.

Features and Benefits

Cerilliant® certified reference materials give your lab accurate, traceable, ready-to-use standards for confident results in clinical, forensic and pharmaceutical testing.

Comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty and traceability

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D203D204F106
description

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

description

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

description

100 μg/mL in methanol, ampule of 0.5 mL, certified reference material, Cerilliant®

description

100 μg/mL in methanol (as free base), ampule of 0.5 mL, certified reference material, Cerilliant®

application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

forensics and toxicology

format

single component solution

format

single component solution

format

single component solution

format

single component solution

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

packaging

ampule of 1 mL

packaging

ampule of 1 mL

packaging

ampule of 0.5 mL

packaging

ampule of 0.5 mL

concentration

100 μg/mL in methanol

concentration

100 μg/mL in methanol

concentration

100 μg/mL in methanol

concentration

-

manufacturer/tradename

Cerilliant®

manufacturer/tradename

Cerilliant®

manufacturer/tradename

Cerilliant®

manufacturer/tradename

-


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

Flash Point (°F)

49.5 °F - closed cup

Flash Point (°C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library